Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with pCR™II-TOPO™ Vector and One Shot™ TOP10 Electrocomp™ E. coli
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
$722.00 - $1282.00
Specifications
Bacterial or Yeast Strain | TOP10 |
---|---|
Cloning Method | TOPO™-TA |
Content And Storage | Box 1: • Topoisomerase I-activated pCR™II-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid. Store at -68 to -85°C. |
Format | Kit |
Product Line | One Shot™ |
Includes: Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid.
Description
Uses a pCR™-TOPO Vector with covalently bound topoisomerase I for fast cloning and recombinants.
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- Choice of T7 (pCR2.1-TOPO) or T7 and SP6 (pCRII-TOPO) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
- EcoR I sites flanking the PCR product insertion site for easy excision of inserts
- Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
- Easy blue/white colony screening for selection of recombinants
Cloning, PCR Cloning
Specifications
TOP10 | |
Box 1: • Topoisomerase I-activated pCR™II-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. Box 2: • One Shot™ Chemically Competent or Electrocomp™ E. coli • S.O.C. medium • Supercoiled pUC19 control plasmid. Store at -68 to -85°C. |
|
One Shot™ | |
T7, SP6 |
TOPO™-TA | |
Kit | |
Cloning Kit | |
TOPO-TA Cloning Vectors |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.