Learn More
Maine Molecular Quality Controls Inc INTROL THROMBOSIS PANEL3X1ML
Supplier: Maine Molecular Quality Controls Inc G1231
INTROL Thrombosis Genotype Panel is intended for in vitro use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of test systems used in the qualitative measurement of the Factor II (Prothrombin), Factor V Leiden and Methylenetetrahydrofolate reductase (MTHFR) genes for mutations Factor II G20210A, Factor V G1691A Leiden, MTHFR C677T and A1298C. The INTROL Thrombosis Genotype Panel consists of three bottles of synthetic DNA of Factor II, Factor V and MTHFR genes suspended in a non-infectious blood-like matrix containing carrier DNA, preservatives and stabilizers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.