Learn More
BioMed Diagnostics For growth and detection of fungi, yeasts, and dermatophytes. | Detects: Trichophyton mentagrophytes, Saccharomyces cerevisiae, Candida albicans, and Aspergillis brasiliensis. | Available in: 20 pack
Supplier: BioMed Diagnostics 11273002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
InTray SAB+PVG is slightly inhibitory against bacteria and is designed for sampling a large spectrum of fungal growth. Bacterial inhibition is further enhanced with the introduction of the antibiotics Polymyxin B, Vancomycin and Gentamicin which is used for samples with suspected high levels of bacterial contamination. | Detects: Trichophyton mentagrophytes, Saccharomyces cerevisiae, Candida albicans, and Aspergillis brasiliensis. | Time to Result: 1 -14 days, incubated at 25C for positive | Available in: 20 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.