Learn More
BioMed Diagnostics Proprietary modified agar that is selective for gonococcal bacteria | Detects: Neisseria gonorrhoeae | Available in: 20 pack
Supplier:  BioMed Diagnostics 11080002
Encompass_Preferred
InTray GC is a fully enclosed microbiology cassette designed to simplify the process of sample collection, transport, culture, and identification of gonococcal bacteria. The proprietary modified Thayer Martin agar is selective for gonococcal bacteria. Results can be interpreted after 24-72 hours of incubation. Suitable for use with oral, rectal, genitourethral, and pharyngeal swabs. | Detects: Neisseria gonorrhoeae | Available in: 20 pack
Catalog No. NC2015890