Learn More
BioMed Diagnostics Highly sensitive and specific chromogenic media for Salmonella spp colonies that are distinguishable by color providing better detection, while inhibiting gram + organisms. | Available in: 20 pack
Supplier:  BioMed Diagnostics 11223002
Encompass_Preferred
Highly sensitive and specific chromogenic media for Salmonella spp colonies that are distinguishable by color providing better detection, while inhibiting gram + organisms. Colorex Salmonella allows for simultaneous growth, detection and chromogenic differentiation of Salmonella species, including S. typhi and S. paratyphi, traditionally taken from clinical stool specimens. Additionally, Colorex Salmonella inhibits the growth of yeasts, mold and fungi resulting in 100% sensitivity and 89% specificity for Salmonella compared to 78% with traditional Hektoen Agar. | Detects: Salmonella enteriditis, Salmonella typhimurium, Eschericia coli, and Citrobacter freundii. | Time to Result: 24 - 48 hours, incubate at 37C | Available in: 20 pack
Catalog No. NC2582925