Learn More
BioMed Diagnostics A selective and chromogenic medium for enterohemorrhagic E. coli O157. | Detects: enterohemorrhagic Eschericia coli O157 | Available in: 20 pack
Supplier:  BioMed Diagnostics 11213002
Encompass_Preferred
A selective and chromogenic medium for enterohemorrhagic E. coli O157. COLOREX O157 is designed for simultaneous growth, observation and chromogenic differentiation of E. coli O157 from clinical or food specimens. This device makes preliminary detection easy by producing distinctive color differences between the growth of E. coli O157 as well as other organisms all while inhibiting the growth of mold, fungi, and other bacteria. | Detects: enterohemorrhagic E. coli O157. | Time to Result: Incubate 18-24 hours at 37C. | Available in: 20 pack
Catalog No. NC2582924