Learn More
BioMed Diagnostics A selective chromogenic medium for the isolation of MRSA. The test can be performed with mixed bacterial populations. | Detects: Methicillin-resistant Staphylococcus aureus | Available in: 20 pack
Supplier: BioMed Diagnostics 11453002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A selective chromogenic medium for the isolation of Staphylococcus aureus (MRSA). The test can be performed with mixed bacterial populations. Intended for identification, not diagnosis. Over recent years, the occurrence of hospital infections caused by methicillin resistant Staphylococcus aureus (MRSA) has been increasing steadily, representing around 20 to 55% of the isolates in Europe and in the USA. Leading cause of nosocomial infections, especially in intensive care units, the MRSA sources are either endogenous (the patient) or through cross contamination (environmental or by person to person contact). | Detects: Methicillin-resistant Staphylococcus aureus | Time to Result: 18 - 24 hours, Incubates at 37C | Available in: 20 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.