Learn More
BioMed Diagnostics A chromogenic medium for use in obtaining a pure culture of gram bacteria. Sub-culture is necessary for identification. | Detects: Carbapenem-resistant Enterobacteria (CRE) | Available in: 20 pack
Supplier: BioMed Diagnostics 11163002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A chromogenic medium for use in obtaining a pure culture of gram bacteria. Sub-culture is necessary for identification. Not intended for diagnosis. | Product Principle: 1.) Combine collection, culture, result and microscopic observation in one IVD device. 2.) Highly selective prepared media in a device designed for in-vitro specimen transport and incubation. 3.) Rapid presumptive-positive diagnosis without re-plating. InTray can be placed directly on microscope stage. 4.) Fully enclosed device with Tyvek air filtration prevents contamination and exposure, post-collection. | Detects: Carbapenem-resistant Enterobacteria (CRE) | Time to Result: 18-24 hours, incubated at 37C for positive | Available in: 20 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.