Learn More
Revvity Health Sciences Inc Insulin standard, 1 Item
Supplier:  Revvity Health Sciences Inc 62INSCDA
Encompass
Purified Insulin can be used as a standard to quantify the Insulin released by cells.
Catalog No. 50-900-644
Explore available promotions