Learn More
Siemens Healthineers INNOVANCE™ Antithrombin Kit
Chromogenic assay for the automated quantitation of functionally active antithrombin in human citrated plasma.
Supplier: Siemens Healthineers 10709521
This item is not returnable.
View return policy
Description
- The INNOVANCE™ Antithrombin assay is an automated, ready-to-use, chromogenic assay that exhibits excellent precision and reliability.
- This quantitative assay for the determination of functional antithrombin utilizes human anti-Xa substrate and avoids interference with heparin cofactor II and direct thrombin inhibitors, such as hirudin.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
The INNOVANCE™ Antithrombin assay is an automated, ready-to-use, chromogenic assay that exhibits excellent precision and reliability. This quantitative assay for the determination of functional antithrombin utilizes human anti-Xa substrate and avoids interference with heparin cofactor II and direct thrombin inhibitors, such as hirudin. | |
Moderate | |
Hemostasis Kit | |
130 Tests/Kit |
Safety and Handling
ShelfLife : 12 Months