Learn More
Restek™ Injection Port Weldments
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For Agilent GCs with OI Purge and Trap Systems
Supplier: Restek™ 22669
Description
- Easily attach a purge and trap with pre-installed, low dead volume fittings
Specifications
GC | |
Split/Splitless | |
Use with OI Purge and Traps | |
Agilent 6890 GCs (With Large Canister Type Filter) |