Learn More
Thermo Scientific ICAP Q QUARTZ TORCH
Supplier: Thermo Scientific 1230790
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Gain improved plasma stability from the tulip design and easy, push-in insertion with this fully demountable, single piece, Thermo Scientific™ Quartz Torch with connection-free gas supply for our Thermo Scientific™ iCAP™ Q ICP-MS system.All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.