Learn More
Siemens Healthineers iCa++ Quick Samplers Kit
For use with Blood Gas/Haemoximetry Analyzers.
Supplier: Siemens Healthineers 11537817
This item is not returnable.
View return policy
Description
The iCa++ Quick™ Sampler is designed for in vitro diagnostic use with lithium heparin tubes, for the measurement of ionized Calcium (iCa++) on RAPIDPoint™ 500 systems and RAPIDLab™ 1260/1265 systems. This is suitable for professional use in a point-of-care or central laboratory environment.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Sampler Kit | |
Blood Gas/Haemoximetry Analyzers |