Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Silencer™ Select Human Long Non-coding RNA (lncRNA) siRNA Library
The Silencer™ Select Human Long Noncoding RNA (lncRNA) siRNA Library contains 15,081 Ambion™ Silencer Select siRNAs targeting 5,027 lncRNAs as identified in GENCODE v22.
$91600.00
Specifications
RNAi Type | siRNA |
---|---|
Species | Human |
Sample Type | RNA |
For Use With (Application) | RNAi |
Product Line | Silencer™Select |
Catalog Number | Mfr. No. | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Price | Quantity & Availability | |||||
A30148
|
Invitrogen™
A30148 |
96-well Plate |
Each of 1 for $91,600.00
|
|
|||||
Description
- Long noncoding RNAs (lncRNAs) have gained interest in recent years as they have been implicated in genomic regulation, cellular housekeeping functions, developmental processes, and disease
- The Silencer™ Select Human Long Noncoding RNA (lncRNA) siRNA Library contains 15,081 Ambion™ Silencer Select siRNAs targeting 5,027 lncRNAs as identified in GENCODE v22
- Library is designed against targeted transcripts aligned to the entire genome, maximizing prediction and coverage across alternatively spliced transcripts
- Silencer Select siRNAs are designed with Thermo Fisher Scientific’s leading design algorithms and locked nucleic acid (LNA) chemical modifications
- This helps ensure efficacy, specificity, and potency to provide a powerful platform for achieving accurate and consistent phenotype data from RNAi screens in the discovery and elucidation of lncRNAs
- Superior efficacy – Reduce off target effects by up to 90% due to latest design algorithms and LNA chemical modifications
- Highest efficiency – Perform knockdown studies with 100-fold more potent siRNAs than currently available siRNAs
- Dependable – Achieve faster discoveries via consistency of gene knockdown and reliability of phenotypic results
- Convenient – Reduce data de-convolution and accelerate target validation with our 96- and 384-well arrayed format
- Customizable – Choose the best option for workflow demands, including additional siRNA scales and pooled formats
- Contains 0.25 nmol of each highly potent siRNA, sufficient for at least 500 transfections (at 5 nM siRNA, 100 μL transfection volume)
Specifications
siRNA | |
RNA | |
Silencer™Select | |
Standard | |
Approved for shipment at Room Temperature or on Wet Ice |
Human | |
RNAi | |
siRNA Library | |
1 Set |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title