Learn More
Bioassay Systems QuantiChrom™ Hydroxyproline Assay Kit. For quantitative determination of hydroxyproline and collagen in biologic and cosmetic samples.
Supplier: Bioassay Systems DHYP100
For quantitative determination of hydroxyproline and collagen in biologic and cosmetic samples. Key Features: Safe. Our hydroxyproline assay uses an improved perchlorate-free chemistry. Sensitive and accurate. Uses 20 µL sample. Linear detection range 0.5 µg/mL to 50 µg/mL hydroxyproline in 96-well plate assay. Convenient. Collagen can be hydrolyzed in Eppendorf tubes, and final incubation can be performed at 37°C. Method: OD560nm. Samples: Hydroxyproline and collagen in biologic and cosmetic samples. Species: All. Procedure: Assay takes Approx. 2 hrs. Kit size: 100 Tests. Detection limit: 0.5 µg/mL.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.