Learn More
Cole-Parmer Stuart SI30H Hybridization Oven; 230 VAC/50 Hz
Supplier: Cole-Parmer 3945405
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Use this hybridization incubator when it is vital to maintain the integrity of the sample, while heating it to the exact temperature for the right period of time. The incubator is ideal for nucleic acid hybridization applications, such as Northern or RNA blots, Southern or DNA blots, and Western or protein blots. This compact incubator comes with two built-in shakers. A rotisserie shaker has a rotational speed from 2 to 10 rpm, while the platform shaker has a rocking speed from 5 to 70 oscillations a minute. The gentle rocking action is ideal for the washing stage of most hybridization procedures. Its safety features include an overtemperature protection system, in which the power is cut to the motor if it overheats or stalls. It has a stainless steel interior for easy cleaning. A double-glazed, smoked acrylic and polycarbonate door provides radiation protection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.