Learn More
Research Products International Corp Hybridization Oven
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 950032
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Unique design features include precise temperature control, speed control and variable bottle positioning. Provides an ideal environment for cDNA library screening, primer synthesis and nucleic acid hybridization. Touch sensitive key pad and microprocessor provide accurate temperature control +/-0.1?C from ambient up to 100?C. An internal fan assists uniform heat distribution throughout the chamber. Rotisserie unit accommodates up to twenty 35 x 150 mm bottles or ten 35 x 300 mm bottles. Combination of both sizes can also be used at the same time. Rotisserie can be replaced with an interchangeable rocking platform. Stainless steel interior allows easy cleaning. Supplied with rotisserie, 6 foot line cord, operating manual describing nucleic acid hybridization techniques and twelve month warranty.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.