Learn More
Research Products International Corp Secure-Seal Hybridization Chambers, Inside Dimensions 20mm Diameter, 25 x 25 Outside Dimensions, 720ul Volume, 40 per Package
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 248870
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Secure-Seal Hybridization Chambers form peel-and-stick enclosures on glass microscope slides. Bond to glass in seconds and remove easily, even after heating. Isolate specimens for hybridization assays, large or multiple specimens, and genomic arrays. Leak-proof chambers are temperature resistant and eliminate evaporation. Chambers provide optimum surface volume for hybridization assays. Minimize friction, promote reagent mixing and facilitate uniform hybridization. Sealable access ports allow addition and removal of reactants.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.