Learn More
Cole-Parmer HI-200-T-120 Hybridization Incubator with Tubes, 80°C; 120 VAC
Supplier: Cole-Parmer 3945508
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
With excellent stability of ±0.1°C within the tubes, these hybridization incubators are great for nucleic hybridization applications such as Northern, Southern, and Western blots.Each incubator features two shakers—rotisserie shaker with rotational speed from 0 to 20 rpm and platform shaker with rocking speed up to 60 oscillation/minute. A removable drip tray provides for easy cleaning. Adjustable feet make leveling on uneven surfaces quick and easy.Please Note: Effective March 2022, the Stuart Hybrigene Incubator Model number HB-3D has now been rebranded Cole-Parmer under Model number of HI-200 and Sku 39455-07. All the same great features and specifications, just with a new and improved look.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.