Learn More
Grace Bio Labs Inc HYBIRDIZATION CHAMBERS SA50
Supplier:  Grace Bio Labs Inc 621502
Encompass_Preferred
SA50-SecureSeal™ 1-13mm Diameter X 0.8mm Depth, 22mm X 25mm OD, 1.5mm Diameter Ports - 120 Port Seals - 50 PACK
Catalog No. NC0814833