Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Helios Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB73091SP
This item is not returnable.
View return policy
Description
Helios Monoclonal specifically detects Helios in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Helios | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
Q9UKS7 | |
IKZF2 | |
E. coli-derived recombinant human Helios Thr3-Gln97 Accession # Q9UKS7 | |
25 μg | |
Primary | |
Detects human Helios in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
736423 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, IKZF2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22807 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Product Content Correction