Learn More
Perkin Elmer US LLC HTS Tubing Accessories Kit
Supplier: Perkin Elmer US LLC N8150144
This Tubing Accessories Kit is designed for use with the HTS in NexION 1000 and 2000 ICP-MS series instruments. It is included in the HTS Peripump Upgrade Kit (N8150141). This kit includes: S10 Sample Probe, 1.0 mm I.D., Qty. 1 (N0812024) ST Nebulizer Line, 0.25 mm I.D., Qty. 1 (N0812026) Meinhard PLUS PFA Capillary F2 Holder, Qty. 1 (N8152716)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.