Learn More
Electron Microscopy Sciences Combination Holder 45° Pin Mount and 90° Profile, Aluminum
Supplier: Electron Microscopy Sciences 7595007A
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For Hitachi S-800, S-4000, S-4100, S-4200, S-4300, S-4500, S-4700 and S-3600N SEMs. For cross sections up to 6.35 mm ( 1/4"). Available in aluminum or brass with stainless steel allen set screws. Includes allen wrench.