Learn More
Thermo Scientific High Pressure Control Region Anion Trap
Supplier: Thermo Scientific 075550
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Anion trap column with anion exchange resin for high pressure 600 psi ion chromatography systems enables continuous regeneration by water to remove anions from eluent and improve conductivity detection by eliminating background contaminants.
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)