Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Hex-Head Fittings for HPLC
SDP

One-piece design, no ferrule required

$73.50 - $221.00

Specifications

Diameter (English) Outer 1/16 in.
Packaging Quantity 10/Pk.
Port Type 10-32
View More Specs

Products 3
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
06-611-693
SDP
View Documents
Restek™
27712
PEEK
Pack of 10 for $100.00
Edge
Add to Cart
 
06-611-694
SDP
View Documents
Restek™
27713
PEEK-HT
Pack of 10 for $221.00
Add to Cart
 
06-611-695
SDP
View Documents
Restek™
27714
PPS
Pack of 10 for $73.50
Add to Cart
 
Description

Description

  • For 1/16 in. tubing on all 10-32 ports
  • Made of biocompatible PEEK
  • Rated to 5,000 psi (350 bar)
Specifications

Specifications

1/16 in.
10/Pk.
10-32
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.