Learn More
Verichem Laboratories HDL LDL CHOLESTEROL VERIF. KIT
Supplier: Verichem Laboratories 9560
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Intended for calibration verification of serum HDL cholesterol and LDL cholesterol. Liquid, ready to use, no manual dilution required. Treat as patient specimen with any direct wet method. Ultra-pure, protein matrix eliminates ambiguity and imprecision, linear relation provided, and assigned values included for AU Chemistry Systems as well as all direct methods.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.