Learn More
AVANTOR MASTERFLEX (VWR LLC) Hand-operated siphon pump, 2 GPM, Polyethylene
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0643900
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Siphon pumps are inexpensive, disposable hand pumps. Model 06439-00 features a squeeze-bulb action while models 06422-20 and 06439-10 feature a bellows action for quick and easy emptying of containers. Model 07079-70 creates a siphon with a few strokes of the pump.