Learn More
Thermo Scientific GUARD COLUMN IONPAX AG9-HC
Supplier: Thermo Scientific 051791
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
IonPac AG9-HC Guard Column (4 x 50 mm), Maximum Operating Pressure: 4000 psi, Mobile Phase Compatibility: pH 0-12; 0-100% HPLC solvents, Bead Diameter: 9.0 micrometer, Pore Size: 2000 angstrom, Crosslinking (%DVB): 55%, Functional Group: Al
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)