Learn More
Thermo Scientific Analytical Column AS9-HC 4x250MM
Supplier: Thermo Scientific 051786
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Analyze trace bromate in drinking water with the Thermo Scientific™ Dionex™ IonPac™ AS9-HC medium-capacity, carbonate eluent, anion-exchange column. Specifically designed for trace bromate in drinking water, the Dionex IonPac AS9-HC column is the specified column for U.S. EPA Methods 300.1 and 317.0. However, the advanced, high-capacity Dionex IonPac AS23 Carbonate Eluent Anion-Exchange column is recommended for determination of oxyhalides and inorganic anions, such as trace bromate, in ozonated drinking water
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.