Learn More
Pfanstiehl Inc L-Histidine, High Purity, Low Endotoxin, Low Metals, USP, EP, BP, JP, ChP (H-116) - 5kg pack GMP, Multicompendial - CAS: 71-00-1
Supplier:  Pfanstiehl Inc H1165KG
Encompass_Preferred
Histidine is commonly used in biotherapeutic formulations as well as bioprocessing. Histidine has been shown to protect monoclonal antibodies in both the liquid and lyophilized states against heat stress and is becoming increasingly common in formulations of protein therapeutics. Histidine is the only amino acid with pH 7.4 within its buffering range, leaving histidine as the best amino acid buffer under physiological conditions. Pfanstiehl High Purity / Low Endotoxin-Low Metal Histidine establishes a new quality standard for amino acids, designed specifically for Biopharmaceutical formulation and bioprocessing. Pfanstiehl Brand Histidine is available as USP (United States Pharmacopoeia), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia).
Catalog No. NC1940139
Explore available promotions