Learn More
Bioassay Systems EnzyChrom™ Glutathione Reductase Kit. For quantitative determination of glutathione reductase activity.
SDP
Supplier:  Bioassay Systems ECGR100
Encompass_Preferred
For quantitative determination of glutathione reductase activity. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.4 to 50 U/L for 20 min reaction. Detection Limit of 0.4 U/L. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD412nm. Samples: Plasma, serum, tissue and culture media etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.4 U/L.
Catalog No. 50-225-5873