Learn More
Cliawaived Inc On Call Pro Blood Glucose Strips (1 box of 50 strips)
Supplier: Cliawaived Inc AC755821
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
On Call Pro Blood Glucose Strips for tesitng glucose in whole blood using the On Call Express Pro and On Call Pro Blood Glucose Monitoring Systems. Contains 50 test strips and a package insert. For Professional use only.