Learn More
Bioassay Systems EnzyChrom™ Glucose Assay Kit III. For quantitative determination of glucose and evaluation of drug effects on glucose metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems EGL3100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of glucose and evaluation of drug effects on glucose metabolism. Key Features: Sensitive and accurate. Use as little as 20 µL samples. Linear detection range in 96-well plate: 0.03 to 2 mM (0.54 mg/dL to 36 mg/dL) glucose. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 30 min at room temperature. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Serum, plasma, urine, saliva, milk, culture medium, food, beverages etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.03 mM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.