Learn More
Andwin Scientific GLASS DISPOSABLE BOX INTMD 6PK
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Andwin Scientific 64411
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Floor Glass Disposable Box features colorful graphics that allow fast identification. This product can be utilized to dispose ampoules, broken glass, vials. The boxes have cardboard outer material with inner 2-mil polypropylene bag. These measure 12 x 12 x 20". It is sold in a pack of 6.