Learn More
B&H PHOTO VIDEO Giottos Rocket Blaster Dust-Removal Tool | Medium | Black
Supplier: B&H PHOTO VIDEO AA1910
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The medium black Giottos Rocket Blaster Dust-Removal Tool safely blows clean air to eliminate dust from sensitive or hard-to-reach surfaces, such as those on lenses, cameras, filters and keyboards. It is crafted from a durable silicone rubber that offers a virtually instantaneous rebound for a nearly continuous stream of clean air. Its long, thin nozzle features an opening designed to eliminate whistling, and can also be removed for cleaning. A one-way valve on the bottom brings in clean air and does not re-distribute dust. Additionally, the blaster stands up on its own and can be set on its side on a flat surface without rolling around.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.