Learn More
Research Products International Corp Gel Drying Frame, 22 x 24 cm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 1076
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Our gel drying frames use the simple and proven technique of natural air drying. Place the gel between two sheets of our highly porous cellophane, clamp gel between frame, then allow to dry overnight. Gels can be preserved almost indefinitely and remain instantly available for photography. This method is recommended for gels up to 1.5mm thick. Each frame set includes corrosion proof stainless steel binders.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.