Learn More
Research Products International Corp Gel Drying Frame, 20 x 36 cm
SDP
Supplier:  Research Products International Corp 1077
Encompass_Preferred
Our gel drying frames use the simple and proven technique of natural air drying. Place the gel between two sheets of our highly porous cellophane, clamp gel between frame, then allow to dry overnight. Gels can be preserved almost indefinitely and remain instantly available for photography. This method is recommended for gels up to 1.5mm thick. Each frame set includes corrosion proof stainless steel binders.
Catalog No. 50-212-133