Learn More
RNA Medical FULL RANGE COOX LVL 3 30X1.2ML
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: RNA Medical QC 2533
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Full Range CO-Oximeter Control, Level 3, 1.2mL, For monitoring the measurements of total hemoglobin and hemoglobin fractions on Co-oximeter instrumentation
This item is not returnable.
View return policy