Learn More
Electron Microscopy Sciences EMS Freezer Hats Planchet A with 2 Chambers
Supplier: Electron Microscopy Sciences 7116701
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Made from brass, these planchets are used for high pressure freezing. They are able to be stacked, allowing for greater depth.
Chamber: 2 Chambers
Depth: 0.1 mm and 0.2 mm