Learn More
Pyramex™ Forestry Kit
Click to view available options
Forestry kit
Description
- Forestry Kit includes Ridgeline Cap Style Hard Hat, S1060 wire mesh face shield, CM6010 Ear Muffs, and HHABCMR Hard Hat Adapter:
- Ridgeline low-profile design features 4-position harness, replaceable sweat band, vented pressure pad, and swinging hinge points
- S1060 mesh face shield provides comfortable air flow and meets ANSI Z87.1 standards
- CM6010 Ear Muffs are dielectric with a NRR of 27 dB
- HHABCMR Hard Hat Adapter conveniently fits into slots and offers pivoting action allowing user to lift face shield while wearing
Specifications
Specifications
ANSI S3.1974 | Yes |
Cushion Color | Black |
For Use With (Equipment) | Kit includes Hard Hat, Faceshield and Hard Hat Adapter |
For Use With (Application) | Forestry |
Model | FORKIT10 |
Pairs per Package | 1 Ea. |
Noise Reduction Rating (NRR) | 27 dB |
Dielectric | Yes |
Style | Hard Hat |