Learn More
Beckman Coulter Aluminum Foil Seals, Package of 100
Supplier: Beckman Coulter 538619
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Lid, Aluminum Foil, Seal and Sample, Bioinert Adhesive Backing, for Deep-Well Titer Plates