Learn More
CTL Scientific Supply Corp Fluoride Test Paper, Box of 200 strips
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp F100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This paper is used for the determination of fluoride ions and gaseous hydrogen fluoride (H2 F2 ). The pinkish-red paper turns yellowish-white in the presence of fluorides. Substantial amounts of chlorates, bromates and sulphates cause white discolorations of the paper. These interferences are readily circumvented. Limit of sensitivity: 20 mg/l fluoride
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.