Learn More
Hillrom FlexiPort™ Reusable Blood Pressure Cuff
Invest in an easy-to-use and affordable reusable blood pressure cuff solution. The single-point FlexiPort snap connection was designed to simplify connectivity, reducing the hassle of needing multiple cuff tubes and connectors. Easily size cuffs up or down for each patient.
$25.00 - $44.40
Specifications
For Use With (Equipment) | Blood Pressure Devices and Patient Monitors |
---|---|
Latex Free | Yes |
Product Type | Blood Pressure Cuff |
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity & Availability | ||||
22-355-156
|
Hillrom
WEL REUSE09 |
09 Child | No Tubes or Connectors |
Each for $25.00
|
|
||||
22-355-157
|
Hillrom
WEL REUSE10 |
10 Small Adult | No Tubes or Connectors |
Each for $25.85
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-355-158
|
Hillrom
WEL REUSE11 |
11 Adult | No Tubes or Connectors |
Each for $26.70
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-355-159
|
Hillrom
WEL REUSE111SC |
11 Adult | 1 Tube, Male Screw Connector |
Each for $26.70
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-355-160
|
Hillrom
WEL REUSE112BV |
11 Adult | 2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve |
Each for $44.40
|
|
||||
22-355-161
|
Hillrom
WEL REUSE122BV |
12 Large Adult | 2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve |
Each for $48.25
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- FlexiPort technology enables every cuff to be used as a one- or two-tube cuff
- Port can be rotated for improved patient comfort
- Materials do not contain natural rubber latex
- Meet all latest clinical guidelines for proper fit from AAMI and AHA
- Primary sizes are color-coded to make it easy to find the proper cuff size
- Up to 13 sizes, ranging from small infant blood pressure cuffs through large adult blood pressure cuffs (including extra-large, thigh and long cuffs)
Specifications
Blood Pressure Devices and Patient Monitors | |
Yes | |
Blood Pressure Cuff |