Learn More
Ace Glass, Inc. Flask, Fernbach, 2800mL, baffled, 38mm OD plain neck, cs/3
Supplier: Ace Glass, Inc. 387531
2800mL baffled Fernbach culture flask with 38mm O.D. long style straight neck for either plastic or stainless steel closures. The bottom of the flask is triple baffled to achieve maximum oxygen transfer to the culture medium. CAUTION: Do not place on direct heat source.