Learn More
PBL Assay Science Fluorescein Isothiocyanate (FITC) Conjugated Anti-Mouse Interferon Alpha (IFN-Alpha) Rat Monoclonal Antibody, Clone RMMA-1 (MAb) [25 micrograms (ug)]
Supplier: PBL Assay Science 221003
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Fluorescein Isothiocyanate (FITC) conjugated rat monoclonal antibody Anti-Mouse Interferon Alpha, Clone RMMA-1 is manufactured by PBL Assay Science (Catalog #22100-3) for research use only (RUO) and packaged in 25 micrograms (ug). This MAb binds to mouse interferon alpha and is suitable for use in flow cytometry and immunohistochemistry applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.