Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Infant Heel Warmers
An easy, safe and reliable way to obtain better heelsticks
This item is not returnable.
View return policy
Description
The increased blood flow makes fewer skin punctures necessary, which allows for less squeezing, fewer infections, less risk of cartilage damage and faster healing.
- Provide safe, nontoxic heat in a format that conforms easily and gently to the infant's heel
- Medical tape allows easy application and removal
- Easy, reliable flex-disk activation
- Available in either liquid or gel
- Consistent 42°C (107°F) temperature provides optimal warming
- “Feather edges” will not scratch infant's foot
Specifications
42°C | |
Flex-disk | |
Infant Heel Warmer | |
Feather edges |
107°F | |
25 Pack | |
Provide safe and nontoxic heat in a format that conforms easily and gently to the infant's heel by using medical tape | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction