Learn More
Fisherbrand™ Applicators
Description
Purified cotton
- Available sterile or nonsterile
- With wooden or plastic handles
Applications:
- Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instruments
Specifications
Plastic | |
Sterile | |
6 in. | |
15cm |
Cotton | |
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments | |
Applicator |