Description
Purified cotton
- Available sterile or nonsterile
- With wooden or plastic handles
Applications:
- Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instruments
Specifications
Autoclavable | |
Cotton | |
15cm | |
Swab |
Wood | |
Non-sterile | |
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments | |
6 in. |