Learn More
Electron Microscopy Sciences FIRM Acrylic Resin Sections 10 ML
Supplier: Electron Microscopy Sciences 1278005
FIRM is a rapid staining technique that provides high-contrast fluorescence microscopy imaging of resin embedded samples, reminiscint of low magnification transmission electron microscopy. FIRM staining is unique in that the brightly fluorescent dye infiltrates the resin, revealing tissue structures in negative relief, with extremely high contrast and resolution. Perfect for previewing specimens prior to EM Analysis. Works with Epoxy and Acrylic type resins.
Simply mount your sections on glass slides, air dry, and overlay with 100 microliters FIRM for 30 seconds, wash and coverslip with water or an aqueous mountant. View in Rhodamine channel of a standard wide field Fluorescence Microscope.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.