Learn More
LabStrong Corp FI-STREEM 40 L STORAGE TANK
Supplier: LabStrong Corp B00121
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Reservoir, Storage; Compatible with FI-Streem III 4L, 8L, and 4L Bi-distiller glass stills; Fully automatic operation, Capacity: 40L; 22D x 18W x 25 in. H , LabStrong model number B00121